Anti-VEGFA Rat Monoclonal Antibody [clone: 124112]

Supplier: R & D Systems
MAB767
220021-529EA 772.25 USD
220021-529
Anti-VEGFA Rat Monoclonal Antibody [clone: 124112]
Antibodies
Type: Primary
Antigen: VEGFA
Clonality: Monoclonal
Clone: 124112
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR