Anti-PLIN3 Mouse Monoclonal Antibody [clone: 779103]

Supplier: R & D Systems
MAB76641
102887-296EA 594.37 USD
102887-296
Anti-PLIN3 Mouse Monoclonal Antibody [clone: 779103]
Antibodies
Type: Primary
Antigen: PLIN3
Clonality: Monoclonal
Clone: 779103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR