Anti-PLIN3 Mouse Monoclonal Antibody [clone: 779119]

Supplier: R & D Systems
MAB7664
102887-294EA 577.02 USD
102887-294
Anti-PLIN3 Mouse Monoclonal Antibody [clone: 779119]
Antibodies
Type: Primary
Antigen: PLIN3
Clonality: Monoclonal
Clone: 779119
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR