Anti-TCF3 Rat Monoclonal Antibody [clone: 826927]

Supplier: R & D Systems
MAB7650
102887-288EA 594.37 USD
102887-288
Anti-TCF3 Rat Monoclonal Antibody [clone: 826927]
Antibodies
Type: Primary
Antigen: TCF3
Clonality: Monoclonal
Clone: 826927
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR