Anti-PSMA2 Mouse Monoclonal Antibody [clone: 797016]

Supplier: R & D Systems
MAB7639
102887-280EA 584.25 USD
102887-280
Anti-PSMA2 Mouse Monoclonal Antibody [clone: 797016]
Antibodies
Type: Primary
Antigen: PSMA2
Clonality: Monoclonal
Clone: 797016
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR