Anti-MILR1 Mouse Monoclonal Antibody [clone: 767727]

Supplier: R & D Systems
MAB7638
102887-278EA 552.43 USD
102887-278
Anti-MILR1 Mouse Monoclonal Antibody [clone: 767727]
Antibodies
Type: Primary
Antigen: MILR1
Clonality: Monoclonal
Clone: 767727
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR