Anti-PSCA Mouse Monoclonal Antibody [clone: 741009]

Supplier: R & D Systems
MAB7636
102887-276EA 594.37 USD
102887-276
Anti-PSCA Mouse Monoclonal Antibody [clone: 741009]
Antibodies
Type: Primary
Antigen: PSCA
Clonality: Monoclonal
Clone: 741009
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR