Anti-LRRC3B Mouse Monoclonal Antibody [clone: 758315]

Supplier: R & D Systems
MAB7635
102887-274EA 577.02 USD
102887-274
Anti-LRRC3B Mouse Monoclonal Antibody [clone: 758315]
Antibodies
Type: Primary
Antigen: LRRC3B
Clonality: Monoclonal
Clone: 758315
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR