Anti-PLIN2 Mouse Monoclonal Antibody [clone: 788620]

Supplier: R & D Systems
MAB7634
102887-272EA 594.37 USD
102887-272
Anti-PLIN2 Mouse Monoclonal Antibody [clone: 788620]
Antibodies
Simple Western

Type: Primary
Antigen: PLIN2
Clonality: Monoclonal
Clone: 788620
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR