Anti-PUM1 Mouse Monoclonal Antibody [clone: 856332]

Supplier: R & D Systems
MAB7628
102887-270EA 561.11 USD
102887-270
Anti-PUM1 Mouse Monoclonal Antibody [clone: 856332]
Antibodies
Type: Primary
Antigen: PUM1
Clonality: Monoclonal
Clone: 856332
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR