Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: 816803]

Supplier: R & D Systems
MAB7606
102887-260EA 594.37 USD
102887-260
Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: 816803]
Antibodies
Simple Western

Type: Primary
Antigen: HSP90AB1
Clonality: Monoclonal
Clone: 816803
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR