Anti-RIPK3 Mouse Monoclonal Antibody [clone: 780115]

Supplier: R & D Systems
MAB7604
102887-258EA 639.2 USD
102887-258
Anti-RIPK3 Mouse Monoclonal Antibody [clone: 780115]
Antibodies
Type: Primary
Antigen: RIPK3
Clonality: Monoclonal
Clone: 780115
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR