Anti-TNFAIP3 Mouse Monoclonal Antibody [clone: 775928]

Supplier: R & D Systems
MAB75981
102887-254EA 577.02 USD
102887-254
Anti-TNFAIP3 Mouse Monoclonal Antibody [clone: 775928]
Antibodies
Type: Primary
Antigen: TNFAIP3
Clonality: Monoclonal
Clone: 775928
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR