Anti-TNFAIP3 Mouse Monoclonal Antibody [clone: 775912]

Supplier: R & D Systems
MAB7598
102887-252EA 594.37 USD
102887-252
Anti-TNFAIP3 Mouse Monoclonal Antibody [clone: 775912]
Antibodies
Type: Primary
Antigen: TNFAIP3
Clonality: Monoclonal
Clone: 775912
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR