Anti-PSMB7 Mouse Monoclonal Antibody [clone: 787807]

Supplier: R & D Systems
MAB7590
102887-250EA 577.02 USD
102887-250
Anti-PSMB7 Mouse Monoclonal Antibody [clone: 787807]
Antibodies
Type: Primary
Antigen: PSMB7
Clonality: Monoclonal
Clone: 787807
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR