Anti-CHRD Rat Monoclonal Antibody [clone: 119721]

Supplier: R & D Systems
MAB758
220021-519EA 763.57 USD
220021-519
Anti-CHRD Rat Monoclonal Antibody [clone: 119721]
Antibodies
Type: Primary
Antigen: CHRD
Clonality: Monoclonal
Clone: 119721
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR