Anti-CD7 Mouse Monoclonal Antibody [clone: 848438]

Supplier: R & D Systems
MAB7579
102887-248EA 600.15 USD
102887-248
Anti-CD7 Mouse Monoclonal Antibody [clone: 848438]
Antibodies
Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: 848438
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR