Anti-SPIB Mouse Monoclonal Antibody [clone: 769653]

Supplier: R & D Systems
MAB7576
102887-244EA 577.02 USD
102887-244
Anti-SPIB Mouse Monoclonal Antibody [clone: 769653]
Antibodies
Type: Primary
Antigen: SPIB
Clonality: Monoclonal
Clone: 769653
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR