Anti-CALD1 Mouse Monoclonal Antibody [clone: 767001]

Supplier: R & D Systems
MAB7569
102887-242EA 566.89 USD
102887-242
Anti-CALD1 Mouse Monoclonal Antibody [clone: 767001]
Antibodies
Type: Primary
Antigen: CALD1
Clonality: Monoclonal
Clone: 767001
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR