Anti-NQO1 Mouse Monoclonal Antibody [clone: 844142]

Supplier: R & D Systems
MAB7567
102887-232EA 561.11 USD
102887-232
Anti-NQO1 Mouse Monoclonal Antibody [clone: 844142]
Antibodies
Type: Primary
Antigen: NQO1
Clonality: Monoclonal
Clone: 844142
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR