Anti-TH Mouse Monoclonal Antibody [clone: 779427]

Supplier: R & D Systems
MAB7566
102887-230EA 647.88 USD
102887-230
Anti-TH Mouse Monoclonal Antibody [clone: 779427]
Antibodies
Simple Western

Type: Primary
Antigen: TH
Clonality: Monoclonal
Clone: 779427
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR