Anti-PSMA1 Mouse Monoclonal Antibody [clone: 785019]

Supplier: R & D Systems
MAB7565
102887-228EA 577.02 USD
102887-228
Anti-PSMA1 Mouse Monoclonal Antibody [clone: 785019]
Antibodies
Type: Primary
Antigen: PSMA1
Clonality: Monoclonal
Clone: 785019
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR