Anti-GLRX3 Mouse Monoclonal Antibody [clone: 791431]

Supplier: R & D Systems
MAB7560
102887-226EA 577.02 USD
102887-226
Anti-GLRX3 Mouse Monoclonal Antibody [clone: 791431]
Antibodies
Type: Primary
Antigen Symbol: GLRX3
Antigen Name: glutaredoxin 3
Antigen Synonyms: GLRX4, GRX3, GRX4, PICOT, TXNL2, TXNL3
Clonality: Monoclonal
Clone: 791431
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR