Anti-CIB1 Mouse Monoclonal Antibody [clone: 791119]

Supplier: R & D Systems
MAB7557
102887-224EA 594.37 USD
102887-224
Anti-CIB1 Mouse Monoclonal Antibody [clone: 791119]
Antibodies
Type: Primary
Antigen: CIB1
Clonality: Monoclonal
Clone: 791119
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR