Anti-TGIF1 Mouse Monoclonal Antibody [clone: 790707]

Supplier: R & D Systems
MAB7555
102887-222EA 577.02 USD
102887-222
Anti-TGIF1 Mouse Monoclonal Antibody [clone: 790707]
Antibodies
Simple Western

Type: Primary
Antigen: TGIF1
Clonality: Monoclonal
Clone: 790707
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR