Anti-NBL1 Rat Monoclonal Antibody [clone: 123224]

Supplier: R & D Systems
MAB7552
220021-515EA 763.57 USD
220021-515
Anti-NBL1 Rat Monoclonal Antibody [clone: 123224]
Antibodies
Type: Primary
Antigen: NBL1
Clonality: Monoclonal
Clone: 123224
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR