Anti-NBL1 Rat Monoclonal Antibody [clone: 123223]

Supplier: R & D Systems
MAB755
220021-513EA 763.57 USD
220021-513
Anti-NBL1 Rat Monoclonal Antibody [clone: 123223]
Antibodies
Type: Primary
Antigen: NBL1
Clonality: Monoclonal
Clone: 123223
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR