Anti-CNTNAP1 Mouse Monoclonal Antibody [clone: 753232]

Supplier: R & D Systems
MAB7548
102887-220EA 594.37 USD
102887-220
Anti-CNTNAP1 Mouse Monoclonal Antibody [clone: 753232]
Antibodies
Type: Primary
Antigen: CNTNAP1
Clonality: Monoclonal
Clone: 753232
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR