Anti-SP7 Mouse Monoclonal Antibody [clone: 764704]

Supplier: R & D Systems
MAB7547
102887-218EA 633.42 USD
102887-218
Anti-SP7 Mouse Monoclonal Antibody [clone: 764704]
Antibodies
Type: Primary
Antigen: SP7
Clonality: Monoclonal
Clone: 764704
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR