Anti-COL4A3 Mouse Monoclonal Antibody [clone: 762102]

Supplier: R & D Systems
MAB7546
102887-216EA 600.15 USD
102887-216
Anti-COL4A3 Mouse Monoclonal Antibody [clone: 762102]
Antibodies
Type: Primary
Antigen: COL4A3
Clonality: Monoclonal
Clone: 762102
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR