Anti-IL4 Mouse Monoclonal Antibody [clone: 140429]

Supplier: R & D Systems
MAB754
220021-511EA 772.25 USD
220021-511
Anti-IL4 Mouse Monoclonal Antibody [clone: 140429]
Antibodies
Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: 140429
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR