Anti-LCOR Mouse Monoclonal Antibody [clone: 731229]

Supplier: R & D Systems
MAB7534
102887-212EA 584.25 USD
102887-212
Anti-LCOR Mouse Monoclonal Antibody [clone: 731229]
Antibodies
Type: Primary
Antigen: LCOR
Clonality: Monoclonal
Clone: 731229
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR