Anti-PGA4 Mouse Monoclonal Antibody [clone: 762015]

Supplier: R & D Systems
MAB7525
102887-206EA 577.02 USD
102887-206
Anti-PGA4 Mouse Monoclonal Antibody [clone: 762015]
Antibodies
Type: Primary
Antigen: PGA4
Clonality: Monoclonal
Clone: 762015
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR