Anti-S1PR3 Mouse Monoclonal Antibody [clone: 776807]

Supplier: R & D Systems
MAB75241
102887-204EA 577.02 USD
102887-204
Anti-S1PR3 Mouse Monoclonal Antibody [clone: 776807]
Antibodies
Type: Primary
Antigen: S1PR3
Clonality: Monoclonal
Clone: 776807
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR