Anti-DIG Mouse Monoclonal Antibody [clone: 611621]

Supplier: R & D Systems
MAB7520
102887-200EA 616.06 USD
102887-200
Anti-DIG Mouse Monoclonal Antibody [clone: 611621]
Antibodies
Type: Primary
Antigen: DIG
Clonality: Monoclonal
Clone: 611621
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR