Anti-GPHN Mouse Monoclonal Antibody [clone: 807423]

Supplier: R & D Systems
MAB7519
102887-198EA 577.02 USD
102887-198
Anti-GPHN Mouse Monoclonal Antibody [clone: 807423]
Antibodies
Type: Primary
Antigen Symbol: GPHN
Antigen Name: gephyrin
Antigen Synonyms: 5730552E08Rik, AI662856, BC027112, C230040D23, GPH, GPHRYN, geph
Clonality: Monoclonal
Clone: 807423
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR