Anti-HSD17B14 Mouse Monoclonal Antibody [clone: 736043]

Supplier: R & D Systems
MAB7515
102887-196EA 561.11 USD
102887-196
Anti-HSD17B14 Mouse Monoclonal Antibody [clone: 736043]
Antibodies
Type: Primary
Antigen: HSD17B14
Clonality: Monoclonal
Clone: 736043
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR