Anti-AHSG Mouse Monoclonal Antibody [clone: 774761]

Supplier: R & D Systems
MAB7510
102887-194EA 577.02 USD
102887-194
Anti-AHSG Mouse Monoclonal Antibody [clone: 774761]
Antibodies
Type: Primary
Antigen: AHSG
Clonality: Monoclonal
Clone: 774761
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR