Anti-CHMP2B Mouse Monoclonal Antibody [clone: 791521]

Supplier: R & D Systems
MAB7509
102887-192EA 577.02 USD
102887-192
Anti-CHMP2B Mouse Monoclonal Antibody [clone: 791521]
Antibodies
Type: Primary
Antigen: CHMP2B
Clonality: Monoclonal
Clone: 791521
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR