Anti-MECOM Mouse Monoclonal Antibody [clone: 702321]

Supplier: R & D Systems
MAB75061
102887-190EA 577.02 USD
102887-190
Anti-MECOM Mouse Monoclonal Antibody [clone: 702321]
Antibodies
Type: Primary
Antigen: MECOM
Clonality: Monoclonal
Clone: 702321
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR