Anti-MECOM Mouse Monoclonal Antibody [clone: 702319]

Supplier: R & D Systems
MAB7506
102887-188EA 577.02 USD
102887-188
Anti-MECOM Mouse Monoclonal Antibody [clone: 702319]
Antibodies
Type: Primary
Antigen: MECOM
Clonality: Monoclonal
Clone: 702319
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR