Anti-TTR Mouse Monoclonal Antibody [clone: 760430]

Supplier: R & D Systems
MAB7505
102887-186EA 633.42 USD
102887-186
Anti-TTR Mouse Monoclonal Antibody [clone: 760430]
Antibodies
Type: Primary
Antigen: TTR
Clonality: Monoclonal
Clone: 760430
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR