Anti-LCK Mouse Monoclonal Antibody [clone: 755103]

Supplier: R & D Systems
MAB7500
102887-182EA 616.06 USD
102887-182
Anti-LCK Mouse Monoclonal Antibody [clone: 755103]
Antibodies
Type: Primary
Antigen: LCK
Clonality: Monoclonal
Clone: 755103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR