Anti-LRIG1 Mouse Monoclonal Antibody [clone: 789211]

Supplier: R & D Systems
MAB7498
102887-180EA 639.2 USD
102887-180
Anti-LRIG1 Mouse Monoclonal Antibody [clone: 789211]
Antibodies
Type: Primary
Antigen: LRIG1
Clonality: Monoclonal
Clone: 789211
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR