Anti-TLR8 Mouse Monoclonal Antibody [clone: 839305]

Supplier: R & D Systems
MAB7496
102887-178EA 594.37 USD
102887-178
Anti-TLR8 Mouse Monoclonal Antibody [clone: 839305]
Antibodies
Simple Western

Type: Primary
Antigen: TLR8
Clonality: Monoclonal
Clone: 839305
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR