Anti-LTA Rat Monoclonal Antibody [clone: 168717]

Supplier: R & D Systems
MAB749
220021-508EA 772.25 USD
220021-508
Anti-LTA Rat Monoclonal Antibody [clone: 168717]
Antibodies
Type: Primary
Antigen: LTA
Clonality: Monoclonal
Clone: 168717
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR