Anti-CD19 Mouse Monoclonal Antibody [clone: 771404]

Supplier: R & D Systems
MAB7489
102886-808EA 594.37 USD
102886-808
Anti-CD19 Mouse Monoclonal Antibody [clone: 771404]
Antibodies
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 771404
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR