Anti-SIRT3 Mouse Monoclonal Antibody [clone: 850911]

Supplier: R & D Systems
MAB7488
102886-806EA 577.02 USD
102886-806
Anti-SIRT3 Mouse Monoclonal Antibody [clone: 850911]
Antibodies
Type: Primary
Antigen: SIRT3
Clonality: Monoclonal
Clone: 850911
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR