Anti-CDH1 Rat Monoclonal Antibody [clone: 114403]

Supplier: R & D Systems
MAB7482
220021-507EA 772.25 USD
220021-507
Anti-CDH1 Rat Monoclonal Antibody [clone: 114403]
Antibodies
Type: Primary
Antigen: CDH1
Clonality: Monoclonal
Clone: 114403
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR