Anti-CDH1 Rat Monoclonal Antibody [clone: 114420]

Supplier: R & D Systems
MAB7481
220021-506EA 594.37 USD
220021-506
Anti-CDH1 Rat Monoclonal Antibody [clone: 114420]
Antibodies
Type: Primary
Antigen: CDH1
Clonality: Monoclonal
Clone: 114420
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR